Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM40L Monoclonal Antibody (OTI5B1), TrueMAB™, OriGene
SDP

Catalog No. 501683565
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3565 Supplier OriGene Supplier No. TA810663
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Potential channel-forming protein implicated in import of protein precursors into mitochondria.

Specifications

Antigen TOMM40L
Applications Western Blot
Classification Monoclonal
Clone OTI5B1
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TOMM40L
Gene Accession No. Q969M1
Gene Alias TOMM40B
Gene Symbols TOMM40L
Host Species Mouse
Immunogen Full length human recombit protein of human TOMM40L produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84134
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.