Learn More
Invitrogen™ TOMM40L Polyclonal Antibody
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (98%), Rat (97%), Rhesus Monkey (99%), Bovine (98%).
Specifications
Specifications
| Antigen | TOMM40L |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TOMM40L |
| Gene Accession No. | Q969M1 |
| Gene Alias | hypothetical protein FLJ12770; mitochondrial import receptor subunit TOM40B; Protein TOMM40-like; RGD1562006; RP11-297K8.10; Tom40B; TOMM40B; Tomm40l; tomm40l {ECO:0000250; translocase of outer mitochondrial membrane 40 homolog (yeast)-like; translocase of outer mitochondrial membrane 40 homolog B; translocase of outer mitochondrial membrane 40 homolog, like; translocase of outer mitochondrial membrane 40 homolog-like; translocase of outer mitochondrial membrane 40 homolog-like (yeast); translocase of outer mitochondrial membrane 40 like; translocase of outer mitochondrial membrane 40B; UniProtKB:Q969M1}; zgc:85715 |
| Gene Symbols | TOMM40L |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.