Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOMM6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552111
Description
Immunogen sequence: MASSTVPVSA AGSANETPEI PDNVGDWLRG VYRFATDRND FRRNLILNLG LFAAGVWLAR NLSDIDLMAP Q Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
TOMM6, also named as OBTP and TOM6, is a part of the preprotein translocase complex of the outer mitochondrial membrane (TOM complex).
Specifications
TOMM6 | |
Polyclonal | |
Unconjugated | |
TOMM6 | |
1110002E23Rik; AI663970; mitochondrial import receptor subunit TOM6 homolog; Obtp; overexpressed breast tumor protein; over-expressed breast tumor protein; Overexpressed breast tumor protein homolog; prickle homolog 4; Prickle4; Tom6; TOMM6; Translocase of outer membrane 6 kDa subunit homolog; translocase of outer mitochondrial membrane 6; translocase of outer mitochondrial membrane 6 homolog; translocase of outer mitochondrial membrane 6 homolog (yeast) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100188893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96B49 | |
TOMM6 | |
Recombinant protein corresponding to Human TOMM6. Recombinant protein control fragment (Product #RP-88758). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction