Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM6 Rabbit anti-Bovine, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502209467
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9467 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9467 Supplier Fabgennix Inc Supplier No. TOM6601AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOMM6 Polyclonal antibody specifically detects TOMM6 in Bovine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Dot Blot, ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen TOMM6
Applications Dot Blot, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene TOMM6
Gene Accession No. Q56JY4, Q96B49, Q9CQN3
Gene Alias 1110002E23Rik; AI663970; mitochondrial import receptor subunit TOM6 homolog; Obtp; overexpressed breast tumor protein; over-expressed breast tumor protein; Overexpressed breast tumor protein homolog; prickle homolog 4; Prickle4; Tom6; TOMM6; Translocase of outer membrane 6 kDa subunit homolog; translocase of outer mitochondrial membrane 6; translocase of outer mitochondrial membrane 6 homolog; translocase of outer mitochondrial membrane 6 homolog (yeast)
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 24-74 on human Mitochondrial import receptor subunit TOM6 homolog.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100188893, 505031, 66119, 681123, 748580
Target Species Bovine, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.