Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Tongs, Stainless Steel with PTFE Tips
SDPSCI_ED

Catalog No. p-200084641
Click to view available options
Sterility:
Non-sterile
Sterile

Safely move items using these tongs with a firm grip.

Stainless steel tongs measure 9 inches long, 3 inches of the tips are coated with PTFE.

  • Available in sterile or non-sterile versions.
  • Autoclavable
  • 304 Stainless Steel
  • PTFE-coated tips

Specifications

Sterility Non-sterile
Material Stainless Steel
Length (English) 22.9 cm
Length (Metric) 9 in.
Type Laboratory Tongs
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.