Learn More
BINDER™ Top Access Port (50 mm)
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series KBF P
Description
- For use on top
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Description | Access port with silicone plug - 50 mm, top |
For Use With (Equipment) | BINDER Chambers Series KBF P |
Material | Steel |
Type | Access Port |
Includes | Silicone plug |
Diameter (Metric) Inside | 50 mm |