Learn More
Vikan™ Top Dog Sleeves, 21 in.
Catalog No. p-7162045
Click to view available options
Color:
Blue
Green
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Sleeves are elastic at both ends to keeps them in place while providing coverage from the wrist to above the elbow.
  • Reusable, 6 mil thick
  • Color-coded
  • FDA-compliant
  • Protect the user from the wrist to above the elbow
  • Elastic at both ends of the sleeves keeps the sleeve securely in place throughout a shift
  • The sleeves work well with Top Dog aprons
  • All sleeves are packaged in pairs
Specifications
Product Type General Purpose Sleeve
Size Universal
Material Ether-Based Polyurethane
Color Red
Sterility Non-sterile
Disposable Reusable
Closure Type Elastic Ends
Seam Type Welded
Cuff Style Elastic
Thickness 6 mil
Certifications/Compliance FDA Compliant
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.