Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Top Grain Pigskin Leather Mig Tig Welding Gloves

Catalog No. p-7230402 Shop All Fisherbrand Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy

Catalog No. 19-065-556A

This item is not returnable. View return policy

Gloves are sewn with Aramid Fiber to provide strength and burn resistance.

  • Top Grain Pigskin leather is smooth and breathable due its porous texture
  • Leather becomes softer with use and withstands moisture without stiffening
  • Stitched with Aramid Fiber for additional strength and burn resistance
  • Split leather gauntlet cuff provides additional forearm protection and allows for easy on/off
  • Straight thumb increases comfort in closed-fisted work
  • Unlined palm for dexterity

Specifications

Cuff Material Split Leather
Color Beige
Material Top Grain Pigskin Leather
Size Small
For Use With (Application) Automotive Assembly, MIG/TIG Welding
Glove Cuff Color Gray
Glove Cuff Style Gauntlet
Series 75-320
Length (English) 11 in.
Length (Metric) 28 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.