Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP1 Mouse anti-Human, Clone: 23B11, Abnova™

Catalog No. 89035582 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-582 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-582 Supplier Abnova Corporation Supplier No. MAB14184

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of human TOP1.

  • High specificity
  • High reactivity

Specifications

Antigen TOP1
Applications Western Blot
Classification Monoclonal
Clone 23B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human TOP1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (PEG, sucrose, 0.09% sodium azide).
Gene Accession No. P11387
Gene Alias TOPI
Gene Symbols TOP1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 699-725 of human TOP1.
Purification Method Size-exclusion chromatography purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7150
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.