Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582658 Supplier Invitrogen™ Supplier No. PA582658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVRASGDAKI KKEKENGFSS PPQIKDEPED DGYFVPPKED IKPLKRPRDE DDADYKPKK Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 92%.

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Top1
Gene Accession No. P11387, Q04750, Q9WUL0
Gene Alias Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase
Gene Symbols Top1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOP1. Recombinant protein control fragment (Product #RP-92579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21969, 64550, 7150
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.