Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP1MT Antibody (TOP1MT/2883R), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308332AF405
Description
TOP1MT Monoclonal antibody specifically detects TOP1MT in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TOP1MT | |
Monoclonal | |
Alexa Fluor 405 | |
mitochondrial, mitochondrial DNA topoisomerase I, topoisomerase (DNA) I, mitochondrial | |
Recombinant full-length human TOP1MT protein (Uniprot: Q969P6) | |
100 μg | |
Cancer | |
116447 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
TOP1MT/2883R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction