Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP186928
Description
TOP2A Polyclonal specifically detects TOP2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TOP2A | |
Polyclonal | |
Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
TOP2A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DASPPKTKTSPKLSNKELKPQKSVVSDLEADDVKGSVPLSSSPPATHFPDETEITNPVPKKNVTVKKTAAKSQSSTSTTGAKKRAAPKGTKRDPALNSGVSQKPDPAKTKNRRKRKPSTSDDSDSNFEKIVSKAVTSKKSKG | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
7153 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction