Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP2A Antibody (TOP2A/6570R), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308757UV
Description
TOP2A Monoclonal antibody specifically detects TOP2A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
TOP2A | |
Monoclonal | |
DyLight 350 | |
DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A | |
Recombinant human TOP2A fragment (aa1352-1493) (exact sequence is proprietary) (Uniprot: P11388 ) | |
100 μg | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
7153 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
TOP2A/6570R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction