Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP2B Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297789R
Description
TOP2B Polyclonal specifically detects TOP2B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TOP2B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
antigen MLAA-44, DNA topoisomerase 2-beta, DNA topoisomerase II beta, DNA topoisomerase II, 180 kD, DNA topoisomerase II, beta isozyme, EC 5.99.1.3, top2beta, TOPIIB, topo II beta, topoisomerase (DNA) II beta (180kD), topoisomerase (DNA) II beta 180kDa, topoisomerase II beta, topoisomerase IIb, U937 associated antigen | |
Produced in rabbits immunized with E. coli-derived Human TOP2B fragment. | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing | |
7155 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction