Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOP2B Monoclonal Antibody (681417)
GREENER_CHOICE

Catalog No. PIMA524310
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524310 Supplier Invitrogen™ Supplier No. MA524310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, approximately 75% cross-reactivity with recombinant mouse TOP2B and no cross-reactivity with recombinant human TOP2A is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

TOP2B (DNA Topoisomerase II Beta) is a Protein Coding gene. This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP2B
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 681417
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TOP2B
Gene Accession No. Q02880, Q64511
Gene Alias antigen MLAA-44; D230016L12Rik; DNA topoisomerase 2-beta; DNA topoisomerase II beta; DNA topoisomerase II, 180 kD; DNA topoisomerase II, beta isozyme; Top-2; Top2b; top2beta; TOPIIB; topo II beta; topoisomerase (DNA) II beta; topoisomerase (DNA) II beta 180kDa; topoisomerase II beta; topoisomerase IIb; U937 associated antigen
Gene Symbols TOP2B
Host Species Mouse
Immunogen E. coli-derived recombinant human TOP2B Phe1192-Asp1621.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21974, 7155
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.