Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOP3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575799
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I (Topo I) and Topo II relax both positive and negative supercoils, Topo III relaxes only negative supercoils. Topo IIIalpha exists as a long and a short isoform, which are produced by alternative splicing. Topo IIIalpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. Topo IIIbeta exists as three isoforms,beta-1,beta -2 and beta-3, also produced by alternative splicing. Topo IIIbeta-1 is expressed in testis, heart, and skeletal muscle, whereas Topo IIIbeta-2 is expressed in thymus, kidney and pancreas.
Specifications
TOP3A | |
Polyclonal | |
Unconjugated | |
Top3a | |
DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; TOP3; TOP3A; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; ZGRF7; zinc finger, GRF-type containing 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
Q13472 | |
Top3a | |
Synthetic peptide corresponding to residues in Human TOP3A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction