Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOP3A Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121538
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121538 100 μL
PIPA5121539 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121538 Supplier Invitrogen™ Supplier No. PA5121538
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I (Topo I) and Topo II relax both positive and negative supercoils, Topo III relaxes only negative supercoils. Topo IIIalpha exists as a long and a short isoform, which are produced by alternative splicing. Topo IIIalpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. Topo IIIbeta exists as three isoforms,beta-1,beta -2 and beta-3, also produced by alternative splicing. Topo IIIbeta-1 is expressed in testis, heart, and skeletal muscle, whereas Topo IIIbeta-2 is expressed in thymus, kidney and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Top3a
Gene Accession No. Q13472
Gene Alias DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; TOP3; TOP3A; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; ZGRF7; zinc finger, GRF-type containing 7
Gene Symbols Top3a
Host Species Rabbit
Immunogen Lys35-Ser179 (Accession Q13472), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.