Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOP3B Polyclonal Antibody
TOP3B Polyclonal Antibody
Supplier: Thermo Scientific PA5101080
Description
TOP3B Polyclonal Antibody for Western Blot, ICC/IF

Specifications
TOP3B | |
Polyclonal | |
Unconjugated | |
TOP3B | |
DNA topoisomerase 3-beta-1; DNA topoisomerase III beta-1; Top3b; Top3b1; Topo III beta; topoisomerase (DNA) III beta; topoisomerase III beta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
21976, 8940 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95985, Q9Z321 | |
TOP3B | |
A synthesized peptide derived from human TOP3B(Accession O95985), corresponding to amino acid residues A817-V862. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction