Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOPAZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561516 Supplier Invitrogen™ Supplier No. PA561516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSASEELKLS CQRTIPMTGK RTWPYYSCAR ISAWCWKKAS LPESSYFLRG SQESCRQVDV PKHQTNQTHL TDSKLLLQSS LTETNTESSS KEKLDSNSNC LS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 59%.

The testis and ovary specific PAZ domain containing 1 (TOPAZ1) protein is a highly conserved protein that is highly expressed in the gonads during germ cell meiosis, suggesting that it may play a role in germ cell development. It contains a PAZ domain and a zinc finger domain and is exclusively expressed in adult testis and fetal ovary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOPAZ1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOPAZ1
Gene Accession No. Q8N9V7
Gene Alias C3orf77; Gm9524; Protein TOPAZ1; RGD1563398; testis and ovary specific PAZ domain containing 1; testis and ovary-specific PAZ domain gene 1; testis- and ovary-specific PAZ domain-containing protein 1; topaz 1; Topaz1
Gene Symbols TOPAZ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOPAZ1. Recombinant protein control fragment (Product #RP-100229).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.