Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOPAZ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562297
Description
Immunogen sequence: KSDTNSIPQL LQTEENVMGV NKLLPEESDL YQSKTNGLLS CLQHEKNKYS IEESSVGRKP RKRMKLSEKA DETVTEMNFS NEYNKSELML QENQM Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 57%.
The testis and ovary specific PAZ domain containing 1 (TOPAZ1) protein is a highly conserved protein that is highly expressed in the gonads during germ cell meiosis, suggesting that it may play a role in germ cell development. It contains a PAZ domain and a zinc finger domain and is exclusively expressed in adult testis and fetal ovary.
Specifications
TOPAZ1 | |
Polyclonal | |
Unconjugated | |
TOPAZ1 | |
C3orf77; Gm9524; Protein TOPAZ1; RGD1563398; testis and ovary specific PAZ domain containing 1; testis and ovary-specific PAZ domain gene 1; testis- and ovary-specific PAZ domain-containing protein 1; topaz 1; Topaz1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9V7 | |
TOPAZ1 | |
Recombinant protein corresponding to Human TOPAZ1. Recombinant protein control fragment (Product #RP-100230). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction