Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOPAZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538060
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538060 Supplier Invitrogen™ Supplier No. PA538060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is human testis tissue lysate. At least three isoforms of TOPAZ1 are known to exist.

The testis and ovary specific PAZ domain containing 1 (TOPAZ1) protein is a highly conserved protein that is highly expressed in the gonads during germ cell meiosis, suggesting that it may play a role in germ cell development. It contains a PAZ domain and a zinc finger domain and is exclusively expressed in adult testis and fetal ovary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOPAZ1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TOPAZ1
Gene Accession No. D3ZUC6, E5FYH1, Q8N9V7
Gene Alias C3orf77; Gm9524; Protein TOPAZ1; RGD1563398; testis and ovary specific PAZ domain containing 1; testis and ovary-specific PAZ domain gene 1; testis- and ovary-specific PAZ domain-containing protein 1; topaz 1; Topaz1
Gene Symbols TOPAZ1
Host Species Rabbit
Immunogen 19 amino acid peptide near the carboxy terminus of human TOPAZ1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301075, 375337, 671232
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.