Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOPBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565178 Supplier Invitrogen™ Supplier No. PA565178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVPDVNTEPSQ NEQIIWDDPT AREERARLAS NLQWPSCPTQ YSELQVDIQN LEDSPFQKPL HDSEIAKQAV CDPGNIRVTE APKHPISEEL ETP Highest antigen sequence identity to the following orthologs - mouse 72%, rat 73%.

This gene encodes a binding protein which interacts with the C-terminal region of topoisomerase II beta. This interaction suggests a supportive role for this protein in the catalytic reactions of topoisomerase II beta through transient breakages of DNA strands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOPBP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOPBP1
Gene Accession No. Q92547
Gene Alias 1110031N14Rik; 2810429C13Rik; AI256758; D430026L04Rik; DNA topoisomerase 2-binding protein 1; DNA topoisomerase II-beta-binding protein 1; DNA topoisomerase II-binding protein 1; KIAA0259; mKIAA0259; RGD1562949; TOP2BP1; TOPBP 1; Topbp1; topoisomerase (DNA) II beta binding protein; topoisomerase (DNA) II binding protein 1
Gene Symbols TOPBP1
Host Species Rabbit
Immunogen Recombinant Human TOPBP1. Recombinant protein control fragment (Product #RP-105926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.