Description
- Lightweight, durable, slotted cap
- Shell constructed of impact-resistant, injection-molded polycarbonate
- Withstands radiant heat loads up to, but not exceeding, 275°F
- 1-Touch™ suspension with one-handed slide adjustment and an easy squeeze opening
- Four-point webbed Fas-Trac™ ratchet suspension with vinyl sweatband and large easy-adjustment knob
Compliance
Third-party ceritfied by SEI to meet Type I, Class E requirements for ANSI/ISEA Z89.1 and CSA Z94.1.
Specifications
Specifications
| Color | White |
| Material | Polycarbonate |
| Suspension Type | Fas-Trac Ratchet |
| Head Style | Non-slotted |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.