Learn More
MSA™ Topgard™ Protective Caps
Catalog No. 189993794 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Red
White
Suspension Type:
1-Touch Suspension
Fas-Trac Ratchet
Head Style:
Non-slotted
Slotted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type Head Style
18-999-3794 White Fas-Trac Ratchet Slotted
18-999-3797 Red Fas-Trac Ratchet Slotted
18-999-3854 Black 1-Touch Suspension Slotted
18-999-3777 White Fas-Trac Ratchet Non-slotted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-999-3794 Supplier MSA™ Supplier No. 475385
Add to Cart
Add to Cart
Designed for use by utilities and for the environments where elevated temperature hazards are present
  • Lightweight, durable, slotted cap
  • Shell constructed of impact-resistant, injection-molded polycarbonate
  • Withstands radiant heat loads up to, but not exceeding, 275°F
  • 1-Touch™ suspension with one-handed slide adjustment and an easy squeeze opening
  • Four-point webbed Fas-Trac™ ratchet suspension with vinyl sweatband and large easy-adjustment knob
Compliance
Third-party ceritfied by SEI to meet Type I, Class E requirements for ANSI/ISEA Z89.1 and CSA Z94.1.
Specifications
Certifications/Compliance Meets ANSI Z89.1-2003; Type I; Class E and CSA Z94.1-2005.
Color White
Material Polycarbonate
Suspension Type Fas-Trac Ratchet
Head Style Slotted
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.