Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Topiramate
GREENER_CHOICE

Catalog No. 362010R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-201-0R 10 mg
36-205-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-201-0R Supplier Tocris Bioscience™ Supplier No. 3620/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GluR5 antagonist; inhibits carbonic anhydrase (CA) II and IV

Topiramate is an anticonvulsant. Antagonizes GluK1 (formerly GluR5) kainate receptors (IC50 = 0.46 μM), acts as a positive allosteric modulator of GABAA receptor-mediated currents, inhibits Nav channels (IC50 = 48.9 μM) and inhibits L-type Ca2+ channels. Also inhibits carbonic anhydrase (CA) (Ki values are 0.1 and 0.2 μM at rat CA II and CA IV respectively), which lowers intracellular neuronal pH.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Specifications

Quantity 10 mg
Product Type Topiramate
Purity >99%
Molecular Weight (g/mol) 339.36
Inhibitors Topiramate

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.