Learn More
Promega TOPK Kinase Enzyme System, 1mg
Catalog No. PRV6568 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6568 1 mg
1 options
Catalog No. PRV6568 Supplier Promega Supplier No. V6568
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human TOPK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TOPK is a MAPK kinase that phosphorylates p38 MAPK and is activated in a cell-cycle-dependent manner in neuronal progenitor cells in vitro.
  • Easily Screen and Profile TOPK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C