Learn More
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, without competent cells
Description
Includes
Topoisomerase I-activated pCR™II-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water
Includes the pCR™II-TOPO TA vector with dual T7 and SP6 promoters. By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable cloning method, featuring a 5-minute cloning reaction and 3-step protocol, and yielding up to 95% recombinants. Resistant to Kanamycin (KanR) andZeocin™ (ZeoR)
Kit Options: TOPO TA Cloning Dual Promoter Kits
TOPO TA Cloning Dual Promoter kits can be purchased with a variety of competent cells that deliver different advantages depending upon your needs:
- General cloning: TOP10 cells (Cat. Nos. K4600-J10, K4600-01, K4600-40)
- High-efficiency cloning: TOP10 Electrocomp™ cells (Cat. Nos. K4660-01, K4660-40)
- General cloning, bacteriophage T1 resistance: DH5α-T1R cells (Cat. Nos. K4620-01, K4620-40)
- Fast growth: Mach1™ -T1R chemically competent E. coli (Cat. No. K4610-20)
- Repressor/induction needs: TOP10F' cells (Cat. Nos. K4650-01, K4650-40)
- Provide your own cells (Cat. Nos. 450640 and 452640)
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- T7 and SP6 promoters for efficient in vitro transcription
- M13 forward and reverse primer sites for sequencing
- 16 convenient restriction sites, including EcoRI, flanking your insert for subsequent excision or subcloning
- Kanamycin and ampicillin resistance for your choice of selection in E. coli
- Easy blue/white colony screening for selection of recombinants
Cloning, PCR Cloning
Specifications
Specifications
| Antibiotic Resistance Bacterial | Ampicillin (AmpR), Kanamycin (KanR) |
| Cloning Method | TOPO™-TA |
| Content And Storage | Box 1: • Topoisomerase I-activated pCR™II-TOPO™ vector • PCR buffer • Salt solution • dNTPs • Control template • M13 forward and reverse primers • Control PCR primers • Sterile water Store at -5 to -30°C. All reagents are stable for 6 months when properly stored. |
| Format | Kit |
| For Use With (Application) | Cloning and subsequent in vitro transcription |
| Includes | Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water |
| Product Line | TOPO™ |
| Product Type | TA Cloning Kit |
| Promoter | T7, SP6 |
| Quantity | 10 Reactions |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.