Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Topoisomerase I Antibody (6D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Topoisomerase I |
---|---|
Clone | 6D8 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
Topoisomerase I Monoclonal antibody specifically detects Topoisomerase I in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
Topoisomerase I | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase | |
A synthesized peptide derived from Human Topoisomerase I [UniProt P11387] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6D8 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7150 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title