Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Topoisomerase II α Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1819S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1819S1 500 μL Concentrated
MS1819S 1 mL Concentrated
MS1819R1 1 mL Prediluted
MS1819R7 7 mL Prediluted
MS1819RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1819S1 Supplier Epredia™ Supplier No. MS1819S1

Topoisomerase II α Ab-4, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Topo II α plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cellsTopo II α is also implicated in drug resistance of tumor cells.

Host Species: Mouse

Clone: Ki-S1

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Lysate of the human histiocytic leukemia cell line U937

Molecular Weight: 170kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Topoisomerase II alpha Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ki-S1
Conjugate Unconjugated
Host Species Mouse
Immunogen Lysate of the human histiocytic leukemia cell line U937
Quantity 500 μL
Regulatory Status IVD
Research Discipline DNA Repair
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.