Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Topoisomerase III alpha Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | Topoisomerase III alpha |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Topoisomerase III alpha Polyclonal antibody specifically detects Topoisomerase III alpha in Mouse samples. It is validated for Western BlotSpecifications
Topoisomerase III alpha | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
DNA topoisomerase 3-alpha, TOP3, topoisomerase (DNA) III alpha | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Topoisomerase III alpha (XP_011522303.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology | |
PBS, 50% glycerol, pH7.3 | |
7156 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title