Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOPORS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TOPORS |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TOPORS Polyclonal specifically detects TOPORS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TOPORS | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
10210 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGATCQIQGVQTNDDLNNDSDDSSDNCVIVGFVKPLAERTPELVELSSDSEDLGSYEKMETVKTQEQEQSYSSGDSDVSRCSSPHSVLGKDEQINKGHCDSSTRIKSKKEEKRSTSLSSPRNL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EC 6.3.2.-, LUNtopoisomerase I binding, arginine/serine-rich, p53-binding protein 3, p53BP3, retinitis pigmentosa 31 (autosomal dominant), RP31, SUMO1-protein E3 ligase Topors, topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase, Topoisomerase I-binding arginine/serine-rich protein, Topoisomerase I-binding RING finger protein, TP53BPLE3 ubiquitin-protein ligase Topors, Tumor suppressor p53-binding protein 3 | |
TOPORS | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title