Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOPORS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TOPORS |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TOPORS Polyclonal specifically detects TOPORS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TOPORS | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q9NS56 | |
10210 | |
This antibody was developed against a recombinant protein corresponding to amino acids: IVQHIIMSNVTRYDLESQAFVSDLRPFLLNRTEHFIHEFISFARSPFNMAAFDQHANYDCPAPSYEEGSHSDSSVITISPDEAETQELDINVATVSQAPWDDETPGPSYSSSEQVHVTMSSLL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 6.3.2.-, LUNtopoisomerase I binding, arginine/serine-rich, p53-binding protein 3, p53BP3, retinitis pigmentosa 31 (autosomal dominant), RP31, SUMO1-protein E3 ligase Topors, topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase, Topoisomerase I-binding arginine/serine-rich protein, Topoisomerase I-binding RING finger protein, TP53BPLE3 ubiquitin-protein ligase Topors, Tumor suppressor p53-binding protein 3 | |
TOPORS | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title