Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOPORS Monoclonal Antibody (4G4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA520967
Description
Peptide Sequence: SPDSKCPICL DRFDNVSYLD RCLHKFCFRC VQEWSKNKAE CPLCKQPFDS IFHSVRAEDD FKEYVLRPSY NGSFVTPDRR FRYRTTLTRE RNASVYSPSG PVNRRTTT.
Functions as an E3 ubiquitin-protein ligase and as an E3 SUMO1-protein ligase. Probable tumor suppressor involved in cell growth, cell proliferation and apoptosis that regulates p53/TP53 stability through ubiquitin-dependent degradation. May regulate chromatin modification through sumoylation of several chromatin modification-associated proteins. May be involved in DNA damage-induced cell death through IKBKE sumoylation.
Specifications
TOPORS | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9NS56 | |
TOPORS | |
TOPORS (NP_005793, 98 a.a. ~ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot, Immunocytochemistry | |
4G4 | |
Unconjugated | |
TOPORS | |
AW105885; E3 ubiquitin-protein ligase Topors; LUN; p53-binding protein 3; P53BP3; p53BP3/LUN; RING-type E3 ubiquitin transferase Topors; RP31; SUMO1-protein E3 ligase Topors; TOP1 binding arginine/serine rich protein; topoisomerase 1-binding RING finger; topoisomerase I binding, arginine/serine-rich; topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase; topoisomerase I-binding arginine/serine-rich protein; Topoisomerase I-binding RING finger protein; Topors; TP53BPL; tumor suppressor p53-binding protein 3 | |
Mouse | |
Affinity chromatography | |
RUO | |
10210 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction