Learn More
Invitrogen™ TOPORS Polyclonal Antibody

Description
Positive Samples: Mouse liver, Mouse lung, Mouse kideny, Rat kidney Immunogen sequence: PDSKCPICLD RFDNVSYLDR CLHKFCFRCV QEWSKNKAEC PLCKQPFDSI FHSVRAEDDF KEYVLRPSYN GSFTNPEVRR FRYRTTMTRE RSASLYSPSS T.
Specifications
Specifications
| Antigen | TOPORS |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TOPORS |
| Gene Accession No. | Q80Z37 |
| Gene Alias | AW105885; E3 ubiquitin-protein ligase Topors; LUN; p53-binding protein 3; P53BP3; p53BP3/LUN; RING-type E3 ubiquitin transferase Topors; RP31; SUMO1-protein E3 ligase Topors; TOP1 binding arginine/serine rich protein; topoisomerase 1-binding RING finger; topoisomerase I binding, arginine/serine-rich; topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase; topoisomerase I-binding arginine/serine-rich protein; Topoisomerase I-binding RING finger protein; Topors; TP53BPL; tumor suppressor p53-binding protein 3 |
| Gene Symbols | TOPORS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.