Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOPORS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564234
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564234 Supplier Invitrogen™ Supplier No. PA564234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVQHIIMSNV TRYDLESQAF VSDLRPFLLN RTEHFIHEFI SFARSPFNMA AFDQHANYDC PAPSYEEGSH SDSSVITISP DEAETQELDI NVATVSQAPW DDETPGPSYS SSEQVHVTMS SLL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Functions as an E3 ubiquitin-protein ligase and as an E3 SUMO1-protein ligase. Probable tumor suppressor involved in cell growth, cell proliferation and apoptosis that regulates p53/TP53 stability through ubiquitin-dependent degradation. May regulate chromatin modification through sumoylation of several chromatin modification-associated proteins. May be involved in DNA damage-induced cell death through IKBKE sumoylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOPORS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOPORS
Gene Accession No. Q9NS56
Gene Alias AW105885; E3 ubiquitin-protein ligase Topors; LUN; p53-binding protein 3; P53BP3; p53BP3/LUN; RING-type E3 ubiquitin transferase Topors; RP31; SUMO1-protein E3 ligase Topors; TOP1 binding arginine/serine rich protein; topoisomerase 1-binding RING finger; topoisomerase I binding, arginine/serine-rich; topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase; topoisomerase I-binding arginine/serine-rich protein; Topoisomerase I-binding RING finger protein; Topors; TP53BPL; tumor suppressor p53-binding protein 3
Gene Symbols TOPORS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOPORS. Recombinant protein control fragment (Product #RP-103605).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.