Learn More
DuPont™ Tyvek™ 400 Pants
GREENER_CHOICE
Catalog No. 19820557 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Tyvek™ 400 Pants come complete with an elastic waist and serged seams.
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • 50 Pants/Cs.
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Tyvek
Color White
Ankle Style Open
Seam Type Serged
Waist Style Elastic
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.