Learn More
LaMotte™ Topsoil Tour Kit
Click to view available options
Product Type:
Topsoil Tour Kit
Description
Includes
Complete lecture materials, illustrated handouts, teacher tips, test procedures, data sheets and games
- Seven units examine and compare the physical and chemical properties of soil
- Tests soil texture, pH, nitrogen, phosphorus and potassium
Order Info
Grades 4 to 8. Class size: 50 students.
Specifications
Specifications
Product Type | Topsoil Tour Kit |
Grade | 4 to 8 |