Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOR1AIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567110 Supplier Invitrogen™ Supplier No. PA567110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGLREPQEDSQ KDLENDPSVN SQAQETTIIA SNAEEAEILH SACGLSKDHQ EVETEGPESA DTGDKSESPD EANVGK Highest antigen sequence identity to the following orthologs - mouse 48%, rat 52%.

One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR1AIP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOR1AIP2
Gene Accession No. Q8NFQ8
Gene Alias 1110020D10Rik; 15 kDa interferon-responsive protein; 15kDa; A130072J07; AA103493; AW060462; AW610675; C77739; Ifrg15; interferon alpha responsive gene, 15 kDa; interferon alpha responsive protein (15 kDa); interferon alpha responsive protein; torsin-1A-interacting protein 2; interferon responsive gene 15; Lull1; lumenal domain like LAP1; lumenal domain-like LAP1; NET9; RP11-12M5.5; TOR1AIP2; torsin 1A interacting protein 2; torsin A interacting protein 2; torsin-1A-interacting protein 2; Torsin-1A-interacting protein 2, isoform IFRG15
Gene Symbols TOR1AIP2
Host Species Rabbit
Immunogen Recombinant Human TOR1AIP2. Recombinant protein control fragment (Product #RP-107753).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.