Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOR1AIP2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989834
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9834 Supplier Bioss Supplier No. BS17326R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

One of the two protein isoforms encoded by this gene is a type II integral membrane protein found in the endoplasmic reticulum (ER). The encoded protein is a cofactor for the ATPase TorsinA, regulating the amount of TorsinA present in the ER compared to that found in the nuclear envelope. Defects in this protein are a cause of early onset primary dystonia, a neuromuscular disease. The other isoform encoded by this gene is an interferon alpha responsive protein whose cellular role has yet to be determined.

Specifications

Antigen TOR1AIP2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene TOR1AIP2
Gene Accession No. Q8NFQ8, Q9H496
Gene Alias 15 kDa interferon-responsive protein; IFRG15; interferon alpha responsive protein; torsin-1A-interacting protein 2; interferon responsive gene 15; LULL1; lumenal domain like LAP1; Lumenal domain-like LAP1; NET9; RP11-12M5.5; TOR1AIP2; torsin 1A interacting protein 2; torsin A interacting protein 2; Torsin-1A-interacting protein 2; Torsin-1A-interacting protein 2, isoform IFRG15
Gene Symbols TOR1AIP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LULL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163590
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.