Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542966 Supplier Invitrogen™ Supplier No. PA542966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VFNNKHSGLW HSGLIDKNLI DYFIPFLPLE YRHVKMCVRA EMRARGSAID Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 79%.

TOR1B may serve as a molecular chaperone assisting in the proper folding of secreted and/or membrane proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TOR1B
Gene Accession No. O14657
Gene Alias 2610016F05Rik; DQ1; FKSG18; TOR1B; Torsin ATPase-1B; torsin B; torsin family 1 member B; torsin family 1, member B; torsin family 1, member B (torsin B); torsin-1B; torsinB
Gene Symbols TOR1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human TOR1B (aa 251-300).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27348
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.