Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553054 Supplier Invitrogen™ Supplier No. PA553054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LITKTALDFW RAGRKREDIQ LKDLEPVLSV GVFNNKHSGL WHSGLIDKNL IDYFIPFLPL EYRHVKMCVR AEMRARGSAI DEDIVTRVAE EMTFFPRDEK IYSDKGCKTV QSRLDFH Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

TOR1B may serve as a molecular chaperone assisting in the proper folding of secreted and/or membrane proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOR1B
Gene Accession No. O14657, Q9ER41
Gene Alias 2610016F05Rik; DQ1; FKSG18; TOR1B; Torsin ATPase-1B; torsin B; torsin family 1 member B; torsin family 1, member B; torsin family 1, member B (torsin B); torsin-1B; torsinB
Gene Symbols TOR1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOR1B. Recombinant protein control fragment (Product #RP-90260).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27348, 30934, 311854
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.