Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOR3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119472
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119472 200 μL
PIPA5121535 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119472 Supplier Invitrogen™ Supplier No. PA5119472
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TOR3A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR3A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TOR3A
Gene Accession No. Q9ER38, Q9H497
Gene Alias Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A
Gene Symbols TOR3A
Host Species Rabbit
Immunogen Recombinant protein Torsin 3A. The antigen corresponds to amino acid range 125-374 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30935, 64222
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.