Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOR3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555972 Supplier Invitrogen™ Supplier No. PA555972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLRGDIINEV VLKLLKAGWS REEITMEHLE PHLQAEIVET IDNGFGHSRL VKENLIDYFI PFLPLEYRHV RLCARDA Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 74%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOR3A
Gene Accession No. Q9H497
Gene Alias Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A
Gene Symbols TOR3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOR3A. Recombinant protein control fragment (Product #RP-94515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.