Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOR3A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$202.00 - $435.00
Specifications
| Antigen | TOR3A |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145371
![]() |
Invitrogen™
PA5145371 |
200 μL |
Each for $435.00
|
|
|||||
|
FEPA5145372
|
Invitrogen™
PA5145372 |
20 μL |
Each for $202.00
|
|
|||||
Description
The function remains unknown.Specifications
| TOR3A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q9ER38 | |
| 30935 | |
| Recombinant Mouse TOR3A protein, Leu111-Asp352 (Accession Q9ER38). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A | |
| TOR3A | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| TOR3A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title