Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOR4A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA560865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560865 Supplier Thermo Scientific Supplier No. PA560865

Rabbit Polyclonal Antibody

Immunogen sequence: SRLVLYPETS RKYRPRVEHR SRAQRCLLLL VAIVGFQVLN AIENLDDNAQ RYDLDGLEKA LQRAVFGQPA AVSRIVALMR D Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

TOR4A gene ontology annotations related to this gene include platelet degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR4A
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOR4A
Gene Accession No. Q9NXH8
Gene Alias C9H9orf167; C9orf167; RGD1308019; TOR4A; Torsin family 4 member A; torsin family 4, member A; torsin family protein C9orf167; torsin-4A
Gene Symbols TOR4A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOR4A
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.