Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Toralizumab Chimeric Recombinant Mouse Monoclonal Antibody (IDEC-131 (Toralizumab))
GREENER_CHOICE

Catalog No. PIMA548309
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548309 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548309 Supplier Invitrogen™ Supplier No. MA548309
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: This antibody is a non agonistic antibody specifc for human CD40 ligand.

Toralizumab works by binding to interferon gamma, which prevents it from interacting with its receptors on immune cells. This blocks the downstream signaling pathways that lead to inflammation and tissue damage in autoimmune diseases. By blocking interferon gamma, toralizumab aims to reduce inflammation and prevent further tissue damage in affected individuals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Toralizumab Chimeric
Applications Inhibition Assays, Immunocytochemistry
Classification Recombinant Monoclonal
Clone IDEC-131 (Toralizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene KRT7
Gene Accession No. P08729
Gene Alias IDEC 131
Host Species Mouse
Immunogen The original monoclonal antibody was generated by immunization with a soluble fusion protein of human gp39 (gp39-CD8).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.