Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TORC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177940
Description
TORC1 Polyclonal specifically detects TORC1 in Human, Mouse, Rat, Bovine, Pufferfish, Zebrafish samples. It is validated for Western Blot, ELISA.Specifications
TORC1 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
CREB regulated transcription coactivator 1, FLJ14027WAMTP1, KIAA0616Transducer of regulated cAMP response element-binding protein 1, MECT1CREB-regulated transcription coactivator 1, mucoepidermoid carcinoma translocated 1, Mucoepidermoid carcinoma translocated protein 1, TORC1TORC-1, Transducer of CREB protein 1, transducer of regulated cAMP response element-binding protein (CREB) 1 | |
Rabbit | |
67.3 kDa | |
0.1 mg | |
Protein Kinase | |
23373 | |
Human, Mouse, Rat, Bovine, Pufferfishes, Zebrafish | |
IgG |
Western Blot, ELISA | |
1 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:10000-1:40000 | |
Q9H8V3 | |
CRTC1 | |
This affinity purified TORC1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 19-34 of human TORC1 protein. (Uniprot: Q6UUV9) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity-purified antibody is directed against human TORC1 protein. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction