Learn More
Invitrogen™ Torc1 Monoclonal Antibody (4G9)

Description
The antibody was affinity-purified from mouse ascites by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | Torc1 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CRTC1 |
| Gene Accession No. | Q6UUV9 |
| Gene Alias | AI413414; CH211-255D18.10; CH211-255D18.9; CREB regulated transcription coactivator 1; CREB regulated transcription coactivator 1b; CREB-regulated transcription coactivator 1; CREB-regulated transcription coactivator 1; CREB-regulated transcription coactivator 1b; CRTC1; crtc1b; FLJ14027; KIAA0616; MECT 1; Mect1; mKIAA0616; mucoepidermoid carcinoma translocated 1; mucoepidermoid carcinoma translocated protein 1; Mucoepidermoid carcinoma translocated protein 1 homolog; R74955; si:ch211-255d18.8; TORC1; TORC-1; transducer of CREB protein 1; transducer of regulated cAMP response element-binding protein (CREB) 1; transducer of regulated cAMP response element-binding protein 1; WAMTP1; zfTorc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.