Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Torc2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583014 Supplier Invitrogen™ Supplier No. PA583014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALNRTSSDSA LHTSVMNPSP QDTYPGPTPP SILPSRRGGI LDGEMDPKVP AIEENLLDDK HLLKPWDAKK LSSSSSRPRS CEVPGIN Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 92%.

CRTC2, also known as TORC2 (Transducers Of Regulated cAMP Response Element-Binding (CREB)) 2 and the related protein CRTC1 are potent CREB coactivators that are exported from the nucleus in a CRM1-dependent manner via phosphorylation-dependent interactions. Studies suggest that their phosphorylation and nuclear/cytoplasmic shuttling play an important role in the regulation of gluconeogenesis by cAMP. CRTC2 is present in both B and T-lymphocytes and abundantly expressed in the thymus. Its activity is important in regulating the expression of genes involved in cellular energy metabolism while CRTC1 is essential for energy balance and fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torc2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRTC2
Gene Accession No. Q53ET0
Gene Alias 4632407F12Rik; CREB regulated transcription coactivator 2; CREB-regulated transcription coactivator 2; CRTC2; mTORC2; OTTHUMP00000035255; RGD1308903; TORC2; TORC-2; transducer of CREB protein 2; transducer of regulated cAMP response element-binding protein (CREB) 2; transducer of regulated cAMP response element-binding protein 2
Gene Symbols CRTC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRTC2. Recombinant protein control fragment (Product #RP-94354).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200186
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.