Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRTC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583679
Description
Immunogen sequence: HGLVERPSRN RFHPLHRRSG DKPGRQFDGS AFGANYSSQP LDESWPRQQP PWKDEKHPGF RLTSALN.
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.
Specifications
CRTC3 | |
Polyclonal | |
Unconjugated | |
Crtc3 | |
2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UUV7 | |
Crtc3 | |
Recombinant protein corresponding to Human CRTC3. Recombinant protein control fragment (Product #RP-100404). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction